Learn More
Sino Biological Recombinant Mouse IL-31 Protein (His Tag)
Supplier:  Sino Biological 51070M08H10
Encompass_Preferred
A DNA sequence encoding the mouse Il31 (NP_083870.1) (Met1-Cys163) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2820