Learn More
Sino Biological Recombinant Mouse R-Spondin 2/RSPO2 Protein (mFc Tag)
Supplier:  Sino Biological 51078M05H50
Encompass_Preferred
A DNA sequence encoding the mouse Rspo2 (NP_766403.1) (Met1-Gly205) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-2830