Learn More
Sino Biological Recombinant Mouse IL-4 Protein (Q136D, Y139D, His Tag)
Supplier:  Sino Biological 51084M08B100
Encompass_Preferred
A DNA sequence encoding the mouse IL4 (P07750) (Met 1-Ser140, Q136D, Y139D) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2833