Learn More
Sino Biological Recombinant Mouse TIE-2 Protein (aa 770-1122, His & GST Tag)
Supplier:  Sino Biological 51087M20B150
Encompass_Preferred
A DNA sequence encoding the mouse TEK (Q02858) (Gln770-Ala1122) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2844