Learn More
Sino Biological Recombinant Mouse IL1R2 Protein (hFc Tag)
Supplier:  Sino Biological 51090M02H100
Encompass_Preferred
A DNA sequence encoding the mouse IL1R2 (P27931) (Met1-Glu355) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2847