Learn More
Sino Biological Recombinant Mouse IL1R2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51090M08H100
Encompass_Preferred
A DNA sequence encoding the mouse IL1R2 (P27931) (Met1-Glu355) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2849