Learn More
Sino Biological Recombinant Mouse EGFR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51091M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2853