Learn More
Sino Biological Recombinant Mouse CNTN5 Protein (His Tag)
Supplier:  Sino Biological 51092M08H50
Encompass_Preferred
A DNA sequence encoding the mouse CNTN5 (P68500) (Met1-Gln1058) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2856