Learn More
Sino Biological Recombinant Mouse ART4/CD297 Protein (hFc Tag)
Supplier:  Sino Biological 51093M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Art4 (NP_080915.1) (Met1-Lys263) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2857