Learn More
Sino Biological Recombinant Mouse ART4/CD297 Protein (His Tag)
Supplier:  Sino Biological 51093M08H100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse ART4 (EDL10575.1) (Met1-Lys 263) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2859