Learn More
Sino Biological Recombinant Mouse IgG2a Protein
Supplier:  Sino Biological 51094MNAH100
Encompass_Preferred
A DNA sequence encoding the Mouse IgG2a Fc region (P01863) (Glu98-Lys330) was expressed and purified.
Catalog No. 50-162-2861