Learn More
Sino Biological Recombinant Mouse IL-17F Protein (His Tag)
Supplier:  Sino Biological 51097M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Il17f (NP_665855.2) (Met1-Ala161) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2869