Learn More
Sino Biological Recombinant Mouse IL-17RB Protein (His Tag)
Supplier:  Sino Biological 51109M08H100
Encompass_Preferred
A DNA sequence encoding the mouse IL17BR (Q9JIP3-1) (Met1-Gly286) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2897