Learn More
Sino Biological Recombinant Mouse CDCP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51110M08H50
Encompass_Preferred
A DNA sequence encoding the mouse CDCP1 (Q5U462) (Met1-Leu666) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2902