Learn More
Sino Biological Recombinant Mouse TAG1/Contactin 2 Protein (His Tag)
Supplier:  Sino Biological 51111M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CNTN2 (Q61330) (Gln31-Glu 1013) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2903