Learn More
Sino Biological Recombinant Mouse MIP-1 alpha/CCL3 Protein
Supplier:  Sino Biological 51114MNAE20
Encompass_Preferred
A DNA sequence encoding the mouse CCL3 (P10855) (Ala24-Ala92) was expressed and purified.
Catalog No. 50-162-2914