Learn More
Sino Biological Recombinant Mouse EphA3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51122M08H200
Encompass_Preferred
A DNA sequence encoding the mouse EPHA3 (NP_034270.1) (Met1-His541) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2921