Learn More
Sino Biological Recombinant Mouse EphA3 Protein (aa 569-984)
Supplier:  Sino Biological 51122MNCB50
Encompass_Preferred
A DNA sequence encoding the mouse EPHA3 (EDK98238.1) (Gly569-Val984) was expressed with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-162-2925