Learn More
Sino Biological Recombinant Mouse IGFBP2 Protein (hFc Tag)
Supplier:  Sino Biological 51124M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Igfbp2 (NP_032368.2) (Met1-Gln305) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2928