Learn More
Sino Biological Recombinant Mouse interferon-alpha/IFNA5 Protein (hFc Tag)
Supplier:  Sino Biological 51126M02H100
Encompass_Preferred
A DNA sequence encoding the mouse IFNA5 (NP_034635.2) (Met1-Glu189) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2936