Learn More
Sino Biological Recombinant Mouse Follistatin-like 1/FSTL1 Protein (hFc Tag)
Supplier:  Sino Biological 51127M02H100
Encompass_Preferred
A DNA sequence encoding the mouse FSTL1 (NP_032073.2) (Met1-Ile306) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2942