Learn More
Sino Biological Recombinant Mouse Follistatin-like 1/FSTL1 Protein (His Tag)
Supplier:  Sino Biological 51127M08H100
Encompass_Preferred
A DNA sequence encoding the mouse FSTL1 (NP_032073.2) (Met1-Ile306) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2944