Learn More
Sino Biological Recombinant Mouse CD70/CD27 Ligand Protein (mFc Tag)
Supplier:  Sino Biological 51129M04H50
Encompass_Preferred
A DNA sequence encoding the mouse Cd70 (NP_035747.1) (Gln47-Pro195) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-162-2951