Learn More
Sino Biological Recombinant Mouse JAML/AMICA Protein (hFc Tag)
Supplier:  Sino Biological 51130M02H100
Encompass_Preferred
A DNA sequence encoding the mouse (NP_001005421.3) (Met1-Leu281) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2952