Learn More
Sino Biological Recombinant Mouse ANTXR2 Protein (hFc Tag)
Supplier:  Sino Biological 51131M02H100
Encompass_Preferred
A DNA sequence encoding the mouse ANTXR2 (NP_598499.1) (Met1-Gly318) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2956