Learn More
Sino Biological Recombinant Mouse ALK4/ACVR1B Protein (hFc Tag)
Supplier:  Sino Biological 51132M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Acvr1b (NP_031421.1) (Met1-Glu126) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2960