Learn More
Sino Biological Recombinant Mouse CD320 Protein (hFc Tag)
Supplier:  Sino Biological 51138M02H100
Encompass_Preferred
A DNA sequence encoding the mouse ICD320 (NP_062294.3) (Met1-Gly208) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2971