Learn More
Sino Biological Recombinant Mouse CD320 Protein (His Tag)
Supplier:  Sino Biological 51138M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD320 (NP_062294.3) (Met1-Gly208) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2973