Learn More
Sino Biological Recombinant Mouse Complement C2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51140M08H20
Encompass_Preferred
A DNA sequence encoding the mouse C2 (NP_038512.2) (Met1-Leu760) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2976