Learn More
Sino Biological Recombinant Mouse CNPY3 Protein (His Tag)
Supplier:  Sino Biological 51146M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Cnpy3 (NP_082341.1) (Met1-Pro272) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2985