Learn More
Sino Biological Recombinant Mouse CRELD2 Protein (hFc Tag)
Supplier:  Sino Biological 51148M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Creld2 (NP_083996.1) (Met1-Leu350) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2991