Learn More
Sino Biological Recombinant Mouse TIM-3/HAVCR2 Protein (hFc Tag)
Supplier:  Sino Biological 51152M02H100
Encompass_Preferred
A DNA sequence encoding the mouse HAVCR2 (AAL65156.1) (Met1-Arg191) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3003