Learn More
Sino Biological Recombinant Mouse CRISP1 Protein (hFc Tag)
Supplier:  Sino Biological 51156M02H50
Encompass_Preferred
A DNA sequence encoding the mouse Crisp1 (NP_033768.3) (Met1-His244) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3012