Learn More
Sino Biological Recombinant Hamster FUT8 Protein (aa 68-575, His Tag)
Supplier:  Sino Biological 51166M08B50
Encompass_Preferred
A DNA sequence encoding the hamster FUT8 (G3HCE4) (Arg68-Lys575) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3028