Learn More
Sino Biological Recombinant Mouse Tnfrsf26 Protein (hFc Tag)
Supplier:  Sino Biological 51169M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Tnfrsf26 (NP_783580.1) (Met1-Lys162) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3029