Learn More
Sino Biological Recombinant Mouse CHL-1/L1CAM-2 Protein
Supplier:  Sino Biological 51176MCCH50
Encompass_Preferred
A DNA sequence encoding the mouse CHL1 (Met1-Gln1027) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-162-3042