Learn More
Sino Biological Recombinant Mouse Siglec-2/CD22 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 51177M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD22 (BAE33829.1)(Met1-Arg708) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3043