Learn More
Sino Biological Recombinant Mouse Siglec-2/CD22 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51177M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD22 (BAE33829.1)(Met1-Arg708) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3045