Learn More
Sino Biological Recombinant Mouse IL-36 gamma/IL-1 F9 Protein (His Tag)
Supplier:  Sino Biological 51179M07E20
Encompass_Preferred
A DNA sequence encoding the mouse IL36g (Q8R460) (Gly13-Ser164) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3048