Learn More
Sino Biological Recombinant Mouse IL-4R Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 51180M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Il4r (NP_001008700.1) (Met1-Arg233) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3049