Learn More
Sino Biological Recombinant Mouse CNTF Protein (His Tag)
Supplier:  Sino Biological 51181M07E20
Encompass_Preferred
A DNA sequence encoding the mouse CNTF (NP_740756.1) (Ala2-Met198) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3052