Learn More
Sino Biological Recombinant Mouse SBDS Protein (His Tag)
Supplier:  Sino Biological 51234M07E100
Encompass_Preferred
A DNA sequence encoding the mouse Sbds (NP_075737.1) (Met1-Glu250) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3074