Learn More
Hach Company Sension+ 5130 Portable Polarographic Dissolved Oxygen Probe
Catalog No. 501997980 Shop All Hach Company Products
Encompass
Sension+ 5130 is a polarographic dissolved oxygen (DO) probe with a durable plastic body, replaceable oxygen-permeable membrane, refillable reference electrolyte, and built-in temperature sensor. The 5130 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable Dissolved Oxygen DO6 meters. The 5130 is ideal for dissolved oxygen measurement in general aqueous applications – particularly in a field environment.Note: Sension+ 5130 DO Probe is compatible with versions of the Sension+ DO6 meter produced since January 2013. To ensure compatibility with your Sension+ DO6 meter, please contact Hach Support for assistance.
Catalog No. 50-199-7980 Supplier Hach Company Supplier No. LZW5130.97.0002
Sension+ 5130 is a polarographic dissolved oxygen (DO) probe with a durable plastic body, replaceable oxygen-permeable membrane, refillable reference electrolyte, and built-in temperature sensor. The 5130 has a fixed 1 meter cable and MP5 connector dedicated for use with Hach Sension+ Portable Dissolved Oxygen DO6 meters. The 5130 is ideal for dissolved oxygen measurement in general aqueous applications – particularly in a field environment.Note: Sension+ 5130 DO Probe is compatible with versions of the Sension+ DO6 meter produced since January 2013. To ensure compatibility with your Sension+ DO6 meter, please contact Hach Support for assistance.