Learn More
Sino Biological Recombinant Mouse IL-28A/Interferon lambda-2 Protein (His Tag)
Supplier:  Sino Biological 51305M08H50
Encompass_Preferred
A DNA sequence encoding the mouse IL28A (NP_001019844.2) (Met1-Val193) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3089