Learn More
Sino Biological Recombinant Mouse IL-28B/Interferon lambda-3 Protein (His Tag)
Supplier:  Sino Biological 51306M08H50
Encompass_Preferred
A DNA sequence encoding the mouse Ifnl3 (NP_796370.1) (Met1-Val193) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3091