Learn More
Hach Company Sension+ 5131 Laboratory Polarographic Dissolved Oxygen Probe
Catalog No. 501997981 Shop All Hach Company Products
Encompass
Sension+ 5131 is a polarographic dissolved oxygen (DO) probe with a durable plastic body, replaceable oxygen-permeable membrane, refillable reference electrolyte, and built-in temperature sensor. The 5131 has a fixed 1 meter cable and BNC + Banana connector dedicated for use with Hach Sension+ Laboratory Dissolved Oxygen MM378 meters. The 5131 is ideal for dissolved oxygen measurement in general aqueous applications.
Catalog No. 50-199-7981 Supplier Hach Company Supplier No. LZW5131.97.0002
Sension+ 5131 is a polarographic dissolved oxygen (DO) probe with a durable plastic body, replaceable oxygen-permeable membrane, refillable reference electrolyte, and built-in temperature sensor. The 5131 has a fixed 1 meter cable and BNC + Banana connector dedicated for use with Hach Sension+ Laboratory Dissolved Oxygen MM378 meters. The 5131 is ideal for dissolved oxygen measurement in general aqueous applications.