Learn More
Sino Biological Recombinant Mouse EphB2 Protein (hFc Tag)
Supplier:  Sino Biological 51367M02H200
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse EPHB2 (NP_034272.1) (Met1-Lys540) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3097