Learn More
Sino Biological Recombinant Mouse MAG/Siglec-4a Protein (ECD, hFc Tag)
Supplier:  Sino Biological 51398M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Mag (NP_034888.1) (Met1-Pro516) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3102