Learn More
Sino Biological Recombinant Mouse MAG/Siglec-4a Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51398M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Mag (NP_034888.1) (Met1-Pro516) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3104