Learn More
Waters Corp Waters Performance Maintenance Kit, For 515 Pump, Annual, HPLC System
Catalog No. 50793643 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 515 pump contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis.
  • Two Clear-100™ plunger seals
  • Two standard sapphire plungers
  • One solvent filter
  • One diffuser
  • Four PerformancePLUS™ Check Valve Cartridges
  • Thin wall Teflon Tubing
Catalog No. 50-793-643 Supplier Waters Corp Supplier No. WAT052587
The Waters™ Performance Maintenance Kit for the 515 pump contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis.
  • Two Clear-100™ plunger seals
  • Two standard sapphire plungers
  • One solvent filter
  • One diffuser
  • Four PerformancePLUS™ Check Valve Cartridges
  • Thin wall Teflon Tubing